1-Phenyl-1,3-butandione ≥98%
Dodavatel: Thermo Scientific
Synonyms:
1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione
A14537.0B
A14537.18
A14537.30
A14537.0BEA
22960
CZK
A14537.0B
A14537.18
A14537.30
1-Phenyl-1,3-butandione ≥98%
1-Phenyl-1,3-butandione
Purity: 98+%
CAS No.: 93-91-4
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 55-60°
Boiling Point: 260-262°
MDL No.: MFCD00008786
Vzorec:
C₆H₅C(O)CH₂C(O)CH₃ MW: 162,19 g/mol Bod varu: 260…262 °C Bod tání: 55…60 °C Hustota: 1,090 Storage Temperature: Ambient |
MDL Number:
MFCD00008786 CAS číslo: 93-91-4 EINECS: 202-286-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...