Tisk…

4,4'-Difluorobenzhydrol ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4,4'-Difluorophenyl phenyl carbinol, 4,4'-Difluorobenzhydryl alcohol, 4,4'-Difluoro-α-phenylbenzyl alcohol, Bis(4-fluorophenyl)carbinol, 4,4'-Difluorodiphenylmethanol, Bis(4-fluorophenyl)methanol

A14370.06 A14370.14 A14370.22
A14370.06EA 676 CZK
A14370.06 A14370.14 A14370.22
4,4'-Difluorobenzhydrol ≥98%
4,4'-Difluorobenzhydrol

Purity: 98+% CAS No.: 365-24-2 Molecular Formula: C13H10F2O Formula Weight: 220.22 Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 142-144°/3mm Beilstein Reference: 2332258 MDL No.: MFCD00000357

Vzorec: C₁₃H₁₀F₂O
Bod varu: 142…144 °C (3 mmHg)
Bod tání: 45…49 °C
Hustota: 1 g/cm³ (20 °C)
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000357
CAS číslo: 365-24-2
EINECS: 206-671-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR