(3-Methoxyphenyl)acetonitrile ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14317.06
A14317.14
A14317.22
A14317.06EA
1260
CZK
A14317.06
A14317.14
A14317.22
(3-Methoxyphenyl)acetonitrile ≥99%
(3-Methoxyphenyl)acetonitrile
Purity: 99%
CAS No.: 19924-43-7
Molecular Formula: C9H9NO
Formula Weight: 147.18
Flash Point: 98°(208°F)
Refractive Index: 1.5312
Boiling Point: 278-282°
Beilstein Reference: 1865539
UN No.: 3276
MDL No.: MFCD00001910
Vzorec:
C₉H₉NO Bod varu: 278…282 °C Hustota: 1,082 Bod vzplanutí: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001910 CAS číslo: 19924-43-7 EINECS: 243-428-5 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...