Tisk…

4-Bromo-3,5-dimethylisoxazole ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

A14179.03 A14179.06 A14179.14
A14179.03EA 1330 CZK
A14179.03 A14179.06 A14179.14
4-Bromo-3,5-dimethylisoxazole ≥97%
4-Bromo-3,5-dimethylisoxazole

Purity: 97% CAS No.: 10558-25-5 Molecular Formula: C5H6BrNO Formula Weight: 176.02 Storage Temperature: 2°C to 8°C Flash Point: 73°(163°F) Refractive Index: 1.4880 Boiling Point: 176° Beilstein Reference: 108522 MDL No.: MFCD00068187

Vzorec: C₅H₆BrNO
Bod varu: 176 °C
Hustota: 1,49
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00068187
CAS číslo: 10558-25-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR