Tisk…

N-Hydroxyphthalimide ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Hydroxyisoindoline-1,3-dione, 2-Hydroxy-1H-isoindole-1,3(2H)-dione, N-Hydroxypthalimide

A13862.22 A13862.36
A13862.22EA 1220 CZK
A13862.22 A13862.36
N-Hydroxyphthalimide ≥98%
N-Hydroxyphthalimide

Purity: 98+% CAS No.: 524-38-9 Molecular Formula: C8H5NO3 Formula Weight: 163.13 Melting Point: ca 233° dec. Beilstein Reference: 131208 Merck Reference: 14,4838 MDL No.: MFCD00005891

Vzorec: C₈H₅NO₃
MW: 163,13 g/mol
Bod varu: 412 °C (1013 hPa)
Bod tání: ∼ 233 °C (rozkl.)
Hustota: 1,58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005891
CAS číslo: 524-38-9
EINECS: 208-358-1
Merck Index: 14,4838

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR