Tisk…

Trimethyl orthoformate ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13760.0F A13760.AE A13760.AP
A13760.0FEA 7420 CZK
A13760.0F A13760.AE A13760.AP
Trimethyl orthoformate ≥99%
Trimethyl orthoformate

Purity: 99% CAS No.: 149-73-5 Molecular Formula: HC(OCH3)3 Formula Weight: 106.12 Flash Point: 15°(59°F) Refractive Index: 1.3790 Melting Point: -53° Boiling Point: 101-102° Beilstein Reference: 969215 Merck Reference: 14,6884 UN No.: 3272 MDL No.: MFCD00008483

Vzorec: C₄H₁₀O₃
MW: 106.12
Bod varu: 101…102 °C
Bod tání: –53 °C
Hustota: 0.968
Bod vzplanutí: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008483
CAS číslo: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3 II
Merck Index: 12,07012

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR