Tisk…

2,4-Dimethylthiazole ≥99%

Dodavatel: Thermo Scientific
Varování

A13604.06 A13604.14 A13604.22
A13604.06EA 1060 CZK
A13604.06 A13604.14 A13604.22
2,4-Dimethylthiazole ≥99%
2,4-Dimethylthiazole

Purity: 99% CAS No.: 541-58-2 Molecular Formula: C5H7NS Formula Weight: 113.18 Flash Point: 42°(107°F) Refractive Index: 1.5090 Odour: Characteristic Boiling Point: 145-147° Solubility: Slightly soluble in water Beilstein Reference: 106414 UN No.: 1993 MDL No.: MFCD00014509

Vzorec: C₅H₇NS
MW: 113,18 g/mol
Bod varu: 145…147 °C
Hustota: 1,057
Bod vzplanutí: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014509
CAS číslo: 541-58-2
EINECS: 208-786-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR