triethylorthoformiát ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A13587.0D
A13587.0F
A13587.AE
A13587.AP
A13587.0DEA
13730
CZK
A13587.0D
A13587.0F
A13587.AE
A13587.AP
triethylorthoformiát ≥98%
triethylorthoformiát
Purity: 98%
CAS No.: 122-51-0
Molecular Formula: C7H16O3
Formula Weight: 148.20
Flash Point: 30°(86°F)
Physical Form: Liquid
Density: 0.891
Refractive Index: 1.3910
Sensitivity: Moisture Sensitive
Appearance: Colourless
Melting Point: -61°
Boiling Point: 144-146°
Beilstein Reference: 605384
Merck Reference: 14,6884
UN No.: 2524
MDL No.: MFCD00009230
Vzorec:
C₇H₁₆O₃ MW: 148.2 Bod varu: 144…146 °C Bod tání: –61 °C Hustota: 0.891 Bod vzplanutí: 30 °C (86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009230 CAS číslo: 122-51-0 EINECS: 204-550-4 UN: 2524 ADR: 3 III Merck Index: 12,07012 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...