Tisk…

2-(4-Methylthiazol-5-yl)ethanol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A13567.14 A13567.22
A13567.14EA 865 CZK
A13567.14 A13567.22
2-(4-Methylthiazol-5-yl)ethanol ≥98%
2-(4-Methylthiazol-5-yl)ethanol

Purity: 98% CAS No.: 137-00-8 Molecular Formula: C6H9NOS Formula Weight: 143.21 Flash Point: 142°(287°F) Refractive Index: 1.5500 Boiling Point: 135-137°/7mm Beilstein Reference: 114249 Merck Reference: 14,6126 MDL No.: MFCD00005339

Vzorec: C₆H₉NOS
Bod varu: 135…137 °C (7 mmHg)
Hustota: 1,196
Bod vzplanutí: 142 °C (287 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005339
CAS číslo: 137-00-8
EINECS: 205-272-6
Merck Index: 14,6126

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR