Tisk…

Heptyl-4-hydroxybenzoate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Heptylparaben, n-Heptyl 4-hydroxybenzoate, Heptyl parahydroxybenzoate, 4-Hydroxybenzoic acid heptyl ester

A13555.14 A13555.22
A13555.14EA 2520 CZK
A13555.14 A13555.22
Heptyl-4-hydroxybenzoate ≥98%
Heptyl-4-hydroxybenzoate

Purity: 98% CAS No.: 1085-12-7 Molecular Formula: C14H20O3 Formula Weight: 236.31 Melting Point: 46-49° MDL No.: MFCD00016481

Vzorec: C₁₄H₂₀O₃
Bod tání: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00016481
CAS číslo: 1085-12-7
EINECS: 214-115-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR