trans-3-Hexenoic acid 97
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13428.09
A13428.18
A13428.09EA
1590
CZK
A13428.09
A13428.18
trans-3-Hexenoic acid 97
trans-3-Hexenoic acid
Purity: 97%
CAS No.: 1577-18-0
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 99°(210°F)
Refractive Index: 1.4398
Odour: Rancid
Melting Point: 11-12°
Boiling Point: 206°
Beilstein Reference: 1700862
UN No.: 3265
MDL No.: MFCD00002786
Vzorec:
C₆H₁₀O₂ MW: 114.14 Bod varu: 206 °C Bod tání: 11…12 °C Hustota: 0.961 Bod vzplanutí: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002786 CAS číslo: 1577-18-0 EINECS: 216-417-8 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...