Propiolic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Acetylenecarboxylic Acid, 2-Propynoic acid
A13245.06
A13245.14
A13245.22
A13245.06EA
1490
CZK
A13245.06
A13245.14
A13245.22
Propiolic acid ≥98%
Propiolic acid
Purity: 98+%
CAS No.: 471-25-0
Molecular Formula: HC≡CCO2H
Formula Weight: 70.05
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.4330
Melting Point: 18°
Boiling Point: 102°/200mm
Beilstein Reference: 878176
Merck Reference: 14,7824
UN No.: 2920
MDL No.: MFCD00004360
Vzorec:
C₃H₂O₂ MW: 70.05 Bod varu: 102 °C (200 mmHg) Bod tání: 18 Hustota: 1.136 Bod vzplanutí: 58 °C (136 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004360 CAS číslo: 471-25-0 EINECS: 207-437-8 UN: 2920 ADR: 8 II Merck Index: 12,08006 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...