(4-Tolyloxy)acetic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-(p-Tolyloxy)acetic acid, 4-Methylphenoxyacetic acid, (4-Methylphenoxy)acetic acid
A13230.09
A13230.18
A13230.30
A13230.09EA
982
CZK
A13230.09
A13230.18
A13230.30
(4-Tolyloxy)acetic acid ≥98%
(4-Tolyloxy)acetic acid
Purity: 98%
CAS No.: 940-64-7
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 137-142°
Beilstein Reference: 2047591
Vzorec:
(C₆H₄(CH₃))OCH₂COOH MW: 166,18 g/mol Bod tání: 137…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014365 CAS číslo: 940-64-7 EINECS: 213-374-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...