Furan ≥99% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm
A13102.AP
A13102.0F
A13102.AE
A13102.APEA
1620
CZK
A13102.AP
A13102.0F
A13102.AE
Furan ≥99% stabilizovaný
Furan
Stabilized with ca 250 ppm BHT
Purity: 99%
CAS No.: 110-00-9
Molecular Formula: C4H4O
Formula Weight: 68.08
Storage Temperature: 2°C to 8°C
Flash Point: -35°(-31°F)
Refractive Index: 1.4220
Melting Point: -86°
Boiling Point: 32-33°
Beilstein Reference: 103221
Merck Reference: 14,4296
UN No.: 2389
MDL No.: MFCD00003222
Vzorec:
C₄H₄O MW: 68,08 g/mol Bod varu: 32…33 °C Bod tání: –86 °C Hustota: 0,936 Bod vzplanutí: –35 °C (–31 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003222 CAS číslo: 110-00-9 EINECS: 203-727-3 UN: 2389 ADR: 3,I Merck Index: 14,4296 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...