(2-Tolyloxy)acetic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-(o-Tolyloxy)acetic acid, (2-Methylphenoxy)acetic acid
A13076.09
A13076.18
A13076.30
A13076.09EA
1550
CZK
A13076.09
A13076.18
A13076.30
(2-Tolyloxy)acetic acid ≥98%
(2-Tolyloxy)acetic acid
Purity: 98%
CAS No.: 1878-49-5
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 152-156°
Beilstein Reference: 1943465
Vzorec:
(C₆H₄CH₃)OCH₂COOH MW: 166.18 Bod tání: 152…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014354 CAS číslo: 1878-49-5 EINECS: 217-517-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...