Tisk…

Trimethylsulphonium iodide ≥99%

Dodavatel: Thermo Scientific
Varování

A12639.0B A12639.18 A12639.30
A12639.0BEA 29100 CZK
A12639.0B A12639.18 A12639.30
Trimethylsulphonium iodide ≥99%
Trimethylsulphonium iodide

Purity: 99% CAS No.: 2181-42-2 Molecular Formula: (CH3)3SI Formula Weight: 204.07 Melting Point: ca 215° subl. Beilstein Reference: 3555192

Vzorec: C₃H₉IS
MW: 204,07 g/mol
Bod tání: ∼ 215 °C (subl.)
Hustota: 1,922 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011632
CAS číslo: 2181-42-2
EINECS: 218-555-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR