Tisk…

Tris(diethylamino)phosphine ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

A12490.06 A12490.14 A12490.22
A12490.06EA 1730 CZK
A12490.06 A12490.14 A12490.22
Tris(diethylamino)phosphine ≥97%
Tris(diethylamino)phosphine

Purity: 97% CAS No.: 2283-11-6 Molecular Formula: [(CH3CH2)2N]3P Formula Weight: 247.37 Flash Point: 58°(136°F) Refractive Index: 1.4737 Boiling Point: 245-246° Beilstein Reference: 636187 UN No.: 1993 MDL No.: MFCD00009041

Vzorec: C₁₂H₃₀N₃P
MW: 247.36
Bod varu: 245…246 °C
Hustota: 0.903
Bod vzplanutí: 58 °C (136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009041
CAS číslo: 2283-11-6
EINECS: 218-920-8
UN: 1993
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR