Kyselina (2,4-Dichlorfenoxy)octová ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,4-D
A12467.0B
A12467.18
A12467.30
A12467.0BEA
10680
CZK
A12467.0B
A12467.18
A12467.30
Kyselina (2,4-Dichlorfenoxy)octová ≥98%
Kyselina (2,4-Dichlorfenoxy)octová
Purity: 98%
CAS No.: 94-75-7
Molecular Formula: C8H6Cl2O3
Formula Weight: 221.04
Odour: Odourless
Melting Point: 136-140°
Boiling Point: 160°/0.4mm
Beilstein Reference: 1214242
Merck Reference: 14,2796
MDL No.: MFCD00004300
Vzorec:
(C₆H₃Cl₂)OCH₂COOH MW: 221,04 g/mol Bod varu: 160 °C (0,4 mmHg) Bod tání: 136…140 °C Hustota: 1,42 Storage Temperature: Ambient |
MDL Number:
MFCD00004300 CAS číslo: 94-75-7 EINECS: 202-361-1 Merck Index: 14,2796 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...