Sodium chloroacetate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Chloroacetic acid sodium salt
A12379.0C
A12379.0E
A12379.30
A12379.36
A12379.0CEA
10680
CZK
A12379.0C
A12379.0E
A12379.30
A12379.36
Sodium chloroacetate ≥98%
Sodium chloroacetate
Purity: 98%
CAS No.: 3926-62-3
Molecular Formula: C2H2ClNaO2
Formula Weight: 116.48
Flash Point: 270°(518°F)
Physical Form: Powder or granules
Odour: Characteristic
pH: 6
Melting Point: ca 199° dec.
Beilstein Reference: 3597157
Merck Reference: 14,2112
UN No.: 2659
MDL No.: MFCD00002684
Vzorec:
C₂H₂ClNaO₂ MW: 116,48 g/mol Bod tání: ∼ 199 °C (rozkl.) Bod vzplanutí: 270 °C (518 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002684 CAS číslo: 3926-62-3 EINECS: 223-498-3 UN: 2659 ADR: 6.1,III Merck Index: 14,2112 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...