Tisk…

3-Fluorophthalic anhydride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Fluoroisobenzofuran-1,3-dione

A12275.06 A12275.14
A12275.06EA 2770 CZK
A12275.06 A12275.14
3-Fluorophthalic anhydride ≥98%
3-Fluorophthalic anhydride

Purity: 98% CAS No.: 652-39-1 Molecular Formula: C8H3FO3 Formula Weight: 166.11 Melting Point: 158-163° Beilstein Reference: 1283580

Vzorec: C₈H₃FO₃
MW: 166,11 g/mol
Bod varu: 351 °C (1013 hPa)
Bod tání: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00039696
CAS číslo: 652-39-1
EINECS: 211-491-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR