2-Aminothiazole 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine
A12026.0E
A12026.18
A12026.22
A12026.30
A12026.36
A12026.0EEA
10200
CZK
A12026.0E
A12026.18
A12026.22
A12026.30
A12026.36
2-Aminothiazole 97%
2-Aminothiazole
Purity: 97%
CAS No.: 96-50-4
Molecular Formula: C3H4N2S
Formula Weight: 100.14
Melting Point: 88-92°
Boiling Point: 117°/15mm
Merck Reference: 14,479
MDL No.: MFCD00005325
Vzorec:
C₃H₄N₂S MW: 100,14 g/mol Bod varu: 117 °C (15 mmHg) Bod tání: 88…92 °C Hustota: 1,5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005325 CAS číslo: 96-50-4 EINECS: 202-511-6 Merck Index: 14,479 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...