(3,4-Dimethoxyphenyl)acetonitrile ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,4-Dimethoxyphenylacetonitrile, Homoveratronitrile, 3,4-Dimethoxybenzyl cyanide
A12003.14
A12003.22
A12003.36
A12003.14EA
817
CZK
A12003.14
A12003.22
A12003.36
(3,4-Dimethoxyphenyl)acetonitrile ≥98%
(3,4-Dimethoxyphenyl)acetonitrile
Purity: 98%
CAS No.: 93-17-4
Molecular Formula: C10H11NO2
Formula Weight: 177.20
Odour: Characteristic
Melting Point: 62-65°
Boiling Point: 177-178°/10mm
Beilstein Reference: 1956100
MDL No.: MFCD00001911
Vzorec:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Bod varu: 177…178 °C (10 mmHg) Bod tání: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001911 CAS číslo: 93-17-4 EINECS: 202-225-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...