Isopropyl cyanoacetate ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Isopropyl 2-cyanoacetate
A11887.14
A11887.22
A11887.14EA
1050
CZK
A11887.14
A11887.22
Isopropyl cyanoacetate ≥97%
Isopropyl cyanoacetate
Purity: 97%
CAS No.: 13361-30-3
Molecular Formula: NCCH2CO2CH(CH3)2
Formula Weight: 127.14
Flash Point: 92°(198°F)
Refractive Index: 1.4150
Boiling Point: 207°
Beilstein Reference: 1756014
MDL No.: MFCD00019842
Vzorec:
C₆H₉NO₂ MW: 127,14 g/mol Bod varu: 207 °C Hustota: 1,013 Bod vzplanutí: 92 °C (198 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019842 CAS číslo: 13361-30-3 EINECS: 236-421-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...