Tisk…

2-Acetyl-5-chlorothiophene ≥99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-(5-Chloro-2-thienyl)-1-ethanone, Rivaroxaban Impurity 20

A11850.09 A11850.18
A11850.09EA 1630 CZK
A11850.09 A11850.18
2-Acetyl-5-chlorothiophene ≥99%
2-Acetyl-5-chlorothiophene

Purity: 99% CAS No.: 6310-09-4 Molecular Formula: C6H5ClOS Formula Weight: 160.62 Flash Point: 108°(226°F) Melting Point: 44-48° Boiling Point: 117-118°/17mm Beilstein Reference: 113936 UN No.: 2811 MDL No.: MFCD00005444

Vzorec: C₆H₅ClOS
MW: 160,62 g/mol
Bod varu: 117…118 °C (17 mmHg)
Bod tání: 44…48 °C
Bod vzplanutí: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005444
CAS číslo: 6310-09-4
EINECS: 228-630-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR