Tisk…

Tin(II) oxalate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Tin (II) ethanedioate, Oxalic acid tin (II) salt

A11689.0E A11689.22 A11689.36
A11689.0EEA 13220 CZK
A11689.0E A11689.22 A11689.36
Tin(II) oxalate ≥98%
Tin(II) oxalate

Purity: 98% CAS No.: 814-94-8 Molecular Formula: C2O4Sn Formula Weight: 206.72 Melting Point: 280° dec. Beilstein Reference: 3708588 Merck Reference: 14,8786 UN No.: 3288 MDL No.: MFCD00040678

Vzorec: C₂O₄Sn
Hustota: 3,56
Storage Temperature: Ambient
MDL Number: MFCD00040678
CAS číslo: 814-94-8
EINECS: 212-414-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,8786

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR