Tisk…

2-Formylbenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Carboxybenzaldehyde, Phthalaldehydic acid

A11201.0B A11201.18 A11201.30
A11201.0BEA 21860 CZK
A11201.0B A11201.18 A11201.30
2-Formylbenzoic acid ≥98%
2-Formylbenzoic acid

Purity: 98+% CAS No.: 119-67-5 Molecular Formula: C8H6O3 Formula Weight: 150.13 Flash Point: 198°(388°F) Melting Point: 97-101° Beilstein Reference: 742381 MDL No.: MFCD00003336

Vzorec: C₈H₆O₃
Bod varu: 101 °C (0,4 mmHg)
Bod tání: 97…101 °C
Hustota: 1,404
Bod vzplanutí: 198 °C (388 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003336
CAS číslo: 119-67-5
EINECS: 204-342-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR