2-Mercapto-5-methylbenzimidazole ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1,3-Dihydro-5-methyl-2H-benzimidazole-2-thione, 2-Mercapto-5-methyl-1H-benzimidazole
A11192.06
A11192.14
A11192.22
A11192.06EA
779
CZK
A11192.06
A11192.14
A11192.22
2-Mercapto-5-methylbenzimidazole ≥98%
2-Mercapto-5-methylbenzimidazole
Purity: 98%
CAS No.: 27231-36-3
Molecular Formula: C8H8N2S
Formula Weight: 164.23
Melting Point: ca 295° dec.
Beilstein Reference: 5083
Vzorec:
C₈H₈N₂S MW: 164,23 g/mol Bod varu: 349 °C (1013 hPa) Bod tání: ∼ 295 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00010617 CAS číslo: 27231-36-3 EINECS: 248-350-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...