Phthalimide potassium salt ≥98%
Dodavatel: Thermo Fisher Scientific
A11134.0E
A11134.22
A11134.36
A11134.0EEA
12840
CZK
A11134.0E
A11134.22
A11134.36
Phthalimide potassium salt ≥98%
Purity: 98+%
CAS No.: 1074-82-4
Molecular Formula: C8H4KNO2
Formula Weight: 185.23
Melting Point:
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...