Tisk…

Ethyl-3,4-dihydroxybenzoate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3,4-Dihydroxybenzoic acid ethyl ester, Protocatechuic acid ethyl ester, Ethyl 3,4-dihydroxybenzoate

A11001.09 A11001.18 A11001.30
A11001.09EA 1800 CZK
A11001.09 A11001.18 A11001.30
Ethyl-3,4-dihydroxybenzoate ≥98%
Ethyl-3,4-dihydroxybenzoate

Purity: 98% CAS No.: 3943-89-3 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 132-135° Beilstein Reference: 2097435

Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Bod tání: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002199
CAS číslo: 3943-89-3
EINECS: 223-529-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR