Tisk…

4,4'-Difluorobenzophenone ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone

A10653.09 A10653.18 A10653.30
A10653.09EA 817 CZK
A10653.09 A10653.18 A10653.30
4,4'-Difluorobenzophenone ≥98%
4,4'-Difluorobenzophenone

Purity: 98+% CAS No.: 345-92-6 Molecular Formula: C13H8F2O Formula Weight: 218.20 Melting Point: 106-110° Boiling Point: 170-172°/10mm MDL No.: MFCD00000353

Vzorec: C₁₃H₈F₂O
Bod varu: 170…172 °C (10 mmHg)
Bod tání: 106…110 °C
Hustota: 1,39 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000353
CAS číslo: 345-92-6
EINECS: 206-466-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR