2-Nitro-1,3,5-trimethoxybenzene ≥98%
Dodavatel: Thermo Scientific
Synonyms:
2,4,6-Trimethoxynitrobenzene, 1,3,5-Trimethoxy-2-nitrobenzene, 1-Nitro-2,4,6-trimethoxybenzene
A10473.06
A10473.14
A10473.22
A10473.06EA
2590
CZK
A10473.06
A10473.14
A10473.22
2-Nitro-1,3,5-trimethoxybenzene ≥98%
2-Nitro-1,3,5-trimethoxybenzene
Purity: 98%
CAS No.: 14227-18-0
Molecular Formula: C9H11NO5
Formula Weight: 213.19
Melting Point: 150-153°
Beilstein Reference: 2277262
Vzorec:
C₉H₁₁NO₅ MW: 213,19 g/mol Bod tání: 150…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016992 CAS číslo: 14227-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...