Benzyltributylammonium chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A10345.0E
A10345.22
A10345.36
A10345.0EEA
12530
CZK
A10345.0E
A10345.22
A10345.36
Benzyltributylammonium chloride ≥98%
Benzyltributylammonium chloride
Purity: 98%
CAS No.: 23616-79-7
Molecular Formula: [CH3(CH2)3]3N(CH2C6H5)Cl
Formula Weight: 311.94
Melting Point: 156-162°
Beilstein Reference: 3776210
Vzorec:
C₁₉H₃₄ClN Bod tání: 156…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011849 CAS číslo: 23616-79-7 EINECS: 245-787-3 UN: 3263 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...