Methyl chloroacetate ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A10325.0E
A10325.30
A10325.0EEA
3370
CZK
A10325.0E
A10325.30
Methyl chloroacetate ≥99%
Methyl chloroacetate
Purity: 99+%
CAS No.: 96-34-4
Molecular Formula: ClCH2CO2CH3
Formula Weight: 108.52
Flash Point: 51°(123°F)
Refractive Index: 1.4220
Melting Point: -32°
Boiling Point: 128-130°
Beilstein Reference: 506255
Merck Reference: 14,6042
UN No.: 2295
MDL No.: MFCD00000931
Vzorec:
C₃H₅ClO₂ MW: 108,52 g/mol Bod varu: 128…130 °C Bod tání: –32 °C Hustota: 1,236 Bod vzplanutí: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000931 CAS číslo: 96-34-4 EINECS: 202-501-1 UN: 2295 ADR: 6.1,I Merck Index: 14,6042 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...