Ethyl isovalerate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester
A10292.0F
A10292.AE
A10292.AP
A10292.0FEA
8620
CZK
A10292.0F
A10292.AE
A10292.AP
Ethyl isovalerate ≥98%
Ethyl isovalerate
Purity: 98%
CAS No.: 108-64-5
Molecular Formula: (CH3)2CHCH2CO2CH2CH3
Formula Weight: 130.19
Flash Point: 26°(78°F)
Refractive Index: 1.3960
Odour: Fruit-like
Melting Point: -99°
Boiling Point: 131-133°
Beilstein Reference: 1744677
Merck Reference: 14,3816
UN No.: 3272
MDL No.: MFCD00009203
Vzorec:
C₇H₁₄O₂ MW: 130,19 g/mol Bod varu: 131…133 °C Bod tání: –99 °C Hustota: 0,866 Bod vzplanutí: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009203 CAS číslo: 108-64-5 EINECS: 203-602-3 UN: 3272 ADR: 3,III Merck Index: 14,3816 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...