Tisk…

Potassium tetrabromoplatinate(II) ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

45548.88 45548.ME
45548.88EA 20390 CZK
45548.88 45548.ME
Potassium tetrabromoplatinate(II) ≥99.9% (metals basis)
Potassium tetrabromoplatinate(II)

Purity: 99.9% CAS No.: 13826-94-3 Molecular Formula: Br4K2Pt Formula Weight: 592.90 Physical Form: Crystalline Melting Point: 300° MDL No.: MFCD00011376

Vzorec: Br₄K₂Pt
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011376
CAS číslo: 13826-94-3
EINECS: 237-536-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR