Tisk…
Dipotassium hexabromopalladate ≥99.999% (metals basis) Pd ≥ 15.5%, Premion®
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: Potassium hexabromopalladate(IV)
43947.03 43947.06
43947.03EA 3510 CZK
43947.03 43947.06
Dipotassium hexabromopalladate ≥99.999% (metals basis) Pd ≥ 15.5%, Premion®
Dipotassium hexabromopalladate
Purity: 99.999% CAS No.: 16919-74-7 Molecular Formula: K2PdBr6 Formula Weight: 664.04 Physical Form: Crystalline
Vzorec: Br₆K₂Pd
MW: 664.04
Storage Temperature: Ambient
MDL Number: MFCD03093651
CAS číslo: 16919-74-7