N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
BSTFA+TMCS (99:1)
43939.06
43939.14
43939.22
43939.46
43939.06EA
1660
CZK
43939.06
43939.14
43939.22
43939.46
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
CAS No.: 25561-30-2
Molecular Formula: C8H18F3NOSi2
Formula Weight: 257.40
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Physical Form: Liquid, Ampouled under argon
Density: 0.97
Refractive Index: 1.3840
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Melting Point: -10°
Boiling Point: 145-147°
Beilstein Reference: 3607620
UN No.: 2924
MDL No.: MFCD00008269
Vzorec:
C₈H₁₈F₃NOSi₂ MW: 257.4 Bod varu: 145…147 °C Bod tání: –10 °C Hustota: 0.97 Bod vzplanutí: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008269 CAS číslo: 25561-30-2 EINECS: 247-103-9 UN: 2924 ADR: 3 III |