Tisk…

1,2-Bis(dimethylsilyl)benzene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1,2-Phenylenebis(dimethylsilane)

43755.09 43755.04
43755.09EA 19800 CZK
43755.09 43755.04
1,2-Bis(dimethylsilyl)benzene ≥98%
1,2-Bis(dimethylsilyl)benzene

Purity: 98% CAS No.: 17985-72-7 Molecular Formula: C10H18Si2 Formula Weight: 194.43 Flash Point: 70°(158°F) Physical Form: Liquid Density: 0.898 Refractive Index: 1.5140 Boiling Point: 101°/13mm MDL No.: MFCD00142462

Vzorec: C₁₀H₁₈Si₂
MW: 194,42 g/mol
Bod varu: 101 °C (13 mmHg)
Hustota: 0,898
Bod vzplanutí: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142462
CAS číslo: 17985-72-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR