Tetraboritan dvojsodný tetrahydrát ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Potassium tetraborate tetrahydrate
39435.36
39435.A3
39435.36EA
1980
CZK
39435.36
39435.A3
Tetraboritan dvojsodný tetrahydrát ≥99%
Tetraboritan dvojsodný tetrahydrát
Purity: 99%
CAS No.: 12045-78-2
Molecular Formula: K2B4O7.4H2O
Formula Weight: 305.51 (233.44anhy)
Physical Form: Granular
Density: 1.74 (anhy)
Melting Point: dec.
Solubility: Soluble in water
Merck Reference: 14,7680
MDL No.: MFCD00149924
Vzorec:
B₄K₂O₇·4H₂O MW: 305.49 Bod tání: Dec. Hustota: 1.74 (anhydr.) Storage Temperature: Ambient |
MDL Number:
MFCD00149924 CAS číslo: 12045-78-2 EINECS: 215-575-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...