Dithizon ≥85% ACS
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Dithizone,for analysis, 1,5-Diphenylthiocarbazone
33283.04
33283.09
33283.18
33283.04EA
858
CZK
33283.04
33283.09
33283.18
Dithizon ≥85% ACS
Dithizon
CAS No.: 60-10-6
Molecular Formula: C13H12N4S
Formula Weight: 256.33
Physical Form: Crystalline
Odour: Odourless
Melting Point: 168° dec.
Beilstein Reference: 748838
Merck Reference: 14,3377
MDL No.: MFCD00003025
Vzorec:
C₆H₅NHNHCSN=NC₆H₅ MW: 256,33 g/mol Bod varu: 406 °C (1013 hPa) Bod tání: 168 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00003025 CAS číslo: 60-10-6 EINECS: 200-454-1 Merck Index: 14,3377 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...