Tisk…

Tetramethylammonium perchlorate

Dodavatel: Thermo Scientific
Nebezpečí

30834.14 30834.22
30834.14EA 4600 CZK
30834.14 30834.22
Tetramethylammonium perchlorate
Tetramethylammonium perchlorate

CAS No.: 2537-36-2 Molecular Formula: (CH3)4NClO4 Formula Weight: 173.60 Physical Form: Crystalline Melting Point: 300° UN No.: 1479 MDL No.: MFCD00043207

Vzorec: C₄H₁₂ClNO₄
MW: 173,6 g/mol
Bod tání: >300 °C
Hustota: 1,379 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00043207
CAS číslo: 2537-36-2
EINECS: 219-805-5
UN: 1479
ADR: 5.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR