Dichloro(methyl)silane ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Methyldichlorosilane
14079.22
14079.36
14079.22EA
2190
CZK
14079.22
14079.36
Dichloro(methyl)silane ≥97%
Dichloro(methyl)silane
Purity: 97+%
CAS No.: 75-54-7
Molecular Formula: CH4Cl2Si
Formula Weight: 115.03
Flash Point: -32°(-26°F)
Physical Form: Liquid
Density: 1.105
Refractive Index: 1.3980
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Melting Point: -93°
Boiling Point: 41-42°
Beilstein Reference: 1071194
UN No.: 1242
MDL No.: MFCD00000494
Vzorec:
CH₄Cl₂Si MW: 115,03 g/mol Bod varu: 41…42 °C Bod tání: –93 °C Hustota: 1,105 Bod vzplanutí: –32 °C (–26 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000494 CAS číslo: 75-54-7 EINECS: 200-877-1 UN: 1242 ADR: 4.3,I REACH: 01-2119436669-21 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...