Chromium hexacarbonyl ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl
13061.03
13061.09
13061.18
13061.03EA
1060
CZK
13061.03
13061.09
13061.18
Chromium hexacarbonyl ≥99%
Chromium hexacarbonyl
Purity: 99%
CAS No.: 13007-92-6
Molecular Formula: C6CrO6
Formula Weight: 220.06
Physical Form: Crystalline
Density: 1.77
Appearance: White
Melting Point: ca 152° dec.
UN No.: 3466
MDL No.: MFCD00010945
Vzorec:
C₆CrO₆ Bod varu: 210 °C (1013 hPa) Hustota: 1,77 Storage Temperature: Ambient |
MDL Number:
MFCD00010945 CAS číslo: 13007-92-6 EINECS: 235-852-4 UN: 3466 ADR: 6.1,III Merck Index: 13,02253 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...