Tisk…

Dipotassium tetrachloropalladate ≥99.99% (metals basis) Pd ≥ 32.2%, Premion®

Dodavatel: Thermo Scientific
Varování

Synonyms: Potassium tetrachloropalladate (II), Potassium chloropalladite

11885.04 11885.09
11885.04EA 7280 CZK
11885.04 11885.09
Dipotassium tetrachloropalladate ≥99.99% (metals basis) Pd ≥ 32.2%, Premion®
Dipotassium tetrachloropalladate

Purity: 99.99% CAS No.: 10025-98-6 Molecular Formula: K2PdCl4 Formula Weight: 326.42 Physical Form: Crystalline Density: 2.67 Appearance: Red-brown Odour: Odourless Melting Point: 105° dec. MDL No.: MFCD00011373

Vzorec: Cl₄K₂Pd
MW: 326,43 g/mol
Bod tání: 105 °C (rozkl.)
Hustota: 2,67
Storage Temperature: Ambient
MDL Number: MFCD00011373
CAS číslo: 10025-98-6
EINECS: 233-049-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR