Tisk…

Dipotassium hexachloropalladate (≥26,3% Pd)

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Potassium hexachloropalladate(IV)

11884.03 11884.06 11884.14
11884.03EA 2960 CZK
11884.03 11884.06 11884.14
Dipotassium hexachloropalladate (≥26,3% Pd)
Dipotassium hexachloropalladate

CAS No.: 16919-73-6 Molecular Formula: K2PdCl6 Formula Weight: 397.32 Physical Form: Crystalline Density: 2.738 Sensitivity: Hygroscopic Appearance: Red Odour: Odourless Melting Point: dec. UN No.: 3077 MDL No.: MFCD00043081

Vzorec: Cl₆K₂Pd
MW: 397,33 g/mol
Bod tání: Dec.
Hustota: 2,738
Storage Temperature: Ambient
MDL Number: MFCD00043081
CAS číslo: 16919-73-6
EINECS: 240-974-6
UN: 3077
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR