Dipotassium tetrabromopalladate (20,0% Pd)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Potassium tetrabromopalladate(II), Potassium tetrabromopalladate (II)
11883.03
11883.06
11883.03EA
3570
CZK
11883.03
11883.06
Dipotassium tetrabromopalladate (20,0% Pd)
Dipotassium tetrabromopalladate
CAS No.: 13826-93-2
Molecular Formula: K2PdBr4
Formula Weight: 504.21
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red-brown
Odour: Odourless
MDL No.: MFCD00049659
Vzorec:
Br₄K₂Pd MW: 504,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049659 CAS číslo: 13826-93-2 EINECS: 237-535-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...