Tisk…

Dysprosium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Dysprosium(III) chloride hydrate

11316.14 11316.22
11316.14EA 3300 CZK
11316.14 11316.22
Dysprosium trichloride hydrate ≥99.9% (REO, rare earth oxide basis)
Dysprosium trichloride hydrate

Purity: 99.9% Molecular Formula: DyCl3.xH2O Formula Weight: 268.86(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic

Vzorec: Cl₃Dy·xH₂O
MW: 268.86 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00010984
EINECS: 233-039-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR