Tisk…

Disodium hexachloroplatinate hexahydrate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Sodium hexachloroplatinate(IV) hexahydrate, Sodium hexachloroplatinate (IV) hexahydrate

11047.03 11047.06
11047.03EA 3750 CZK
11047.03 11047.06
Disodium hexachloroplatinate hexahydrate
Disodium hexachloroplatinate hexahydrate

CAS No.: 19583-77-8 Molecular Formula: Na2PtCl6.6H2O Formula Weight: 561.88 (453.79anhy) Physical Form: Crystalline Density: 2.50 Appearance: Orange Odour: Odourless Melting Point: 110° -6H2O Solubility: Soluble in water, alcohol. Insoluble in ether Merck Reference: 14,8623 UN No.: 3288 MDL No.: MFCD00149172


Catalyst

Vzorec: Cl₆Na₂Pt·6H₂O
Bod tání: 110 °C (-6H₂O)
Hustota: 2,50
Storage Temperature: Ambient
MDL Number: MFCD00149172
CAS číslo: 19583-77-8
EINECS: 240-983-5
UN: 3288
ADR: 6.1,III
Merck Index: 14,8623

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR