Tisk…

Tris(diethylamino)phosphine 95%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Hexaethyltriaminophosphine, Hexaethylphosphorus triamide, Hexaethyl phosphorous triamide

423080250 423080050 423081000
ACRO423080250EA 6620 CZK
ACRO423080250 ACRO423080050 ACRO423081000
Tris(diethylamino)phosphine 95%
Tris(diethylamino)phosphine
Vzorec: C₁₂H₃₀N₃P
MW: 247.36
Bod varu: 111 °C (6 mmHg)
Hustota: 0.89 g/cm³
Bod vzplanutí: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00009041
CAS číslo: 2283-11-6
EINECS: 218-920-8
UN: 1993
ADR: 3 III

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow to light orange Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR