Tisk…

(4-Hydroxyphenoxy)acetic acid 98%

Dodavatel: Thermo Scientific

412090250 412090050
ACRO412090250EA 5590 CZK
ACRO412090250 ACRO412090050
(4-Hydroxyphenoxy)acetic acid 98%
(4-Hydroxyphenoxy)acetic acid
Vzorec: (C₆H₄(OH))OCH₂COOH
MW: 168,15 g/mol
MDL Number: MFCD00014362
CAS číslo: 1878-84-8

Objednat

Výsledky specifikačního testu

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 152 to 156 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR